Organic reference standard, Anthracene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA20520000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:120-12-7
- Description:Anthracene 100 µg/ml in acetonitrile
- Formula:C₁₄H₁₀
- Description:Anthracene