
Ruthenium(IV) oxide hydrate (> 54% Ru)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32740-79-7
:
Formula:O₂Ru·H₂O
MDL Number:MFCD00149846
:151.08 g/mol
Density:6,97 g/cm³ (20 °C)
EINECS:234-840-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Black Powder
- Assay:≥54 % (Ru)