



Rhodium(III) chloride hydrate 38% Rh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20765-98-4
:
Formula:Cl₃Rh·H₂O
MDL Number:MFCD00149839
:227.28 g/mol
Boiling Pt:800 °C (1013 hPa)
Melting Pt:100 °C (dec.)
Density:Min, 1 g/cm³ (20 °C)
EINECS:233-165-4
UN:3260
:GHS07
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Red-brown to dark brown Crystalline powder
- Assay:37 to 40 % (Rh)