

Manganese(II) chloride tetrahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:13446-34-9
:
Formula:MnCl₂·4H₂O
MDL Number:MFCD00149792
:197.9 g/mol
Merck Index:13,05751
Boiling Pt:1190 °C (1013 hPa)
Melting Pt:58…59 °C
Density:2.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-869-6
UN:3077
ADR:9,III
:GHS07
:GHS09