Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Talc
Talc
Talc
ROTH9486.1
:
Roth Carl
:
Talc
ROTH9486.1
:
Roth Carl
:
Pack type:
Glass bottle for solids
Pk:
2,5 kg
:
:
379.27 g/mol
:
13,09127
:
>1200 °C
:
2.58…2.83 g/cm³ (20 °C)
:
238-877-9
:
GHS07
:
Ambient
:
14807-96-6
:
Mg₃O₁₁Si₄·1H₂O
:
MFCD00792903