

Barium hydroxide octahydrate ≥95%, TECHNICAL
Catalog # 21678.515
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
Specifications
Specification Test Results
- Assay:Min. 95 %
Specification Test Results
About this item
:
:315.47 g/mol
Merck Index:13,00980
Boiling Pt:780 °C (1013 hPa)
Melting Pt:78 °C
Density:2.18 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:241-234-5
UN:3262
ADR:8,III
:GHS05
:GHS07
CAS Number:12230-71-6
Formula:Ba(OH)₂·8H₂O
MDL Number:MFCD00149152