Order Entry
Export
ContactUsLinkComponent
Sodium sulphate decahydrate
Sodium sulphate decahydrate
Catalog # ROTHT108.1
Supplier:  Roth Carl
CAS Number:  
Sodium sulphate decahydrate
Catalog # ROTHT108.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

About this item

 :
 :322.2 g/mol
Merck Index:13,08755
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:32.4 °C
Density:1.464 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7727-73-3
Formula:Na₂SO₄·10H₂O
MDL Number:MFCD00149189