Sodium thiosulphate pentahydrate, Sigma-Aldrich®
Catalog # SIAL380016-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:248.19 g/mol
Merck Index:12,08844
Melting Pt:48 °C
Density:1.67 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:10102-17-7
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186