

tetra-Chloroauric(III) acid trihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16961-25-4
:
Formula:HAuCl₄·3H₂O
MDL Number:MFCD00149904
:393.83 g/mol
Melting Pt:30 °C
Density:3.9 g/cm³ (20 °C)
EINECS:240-948-4
UN:3260
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
Storage Temperature:Ambient