

tetra-Chloroauric(III) acid trihydrate
Catalog # ICNA0215252601
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:393.83 g/mol
Melting Pt:30 °C
Density:3.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-948-4
UN:3260
ADR:8,III
:GHS05
:GHS07
CAS Number:16961-25-4
Formula:HAuCl₄·3H₂O
MDL Number:MFCD00149904