Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
p-Toluenesulphonic acid monohydrate
p-Toluenesulphonic acid monohydrate, Sigma-Aldrich®
p-Toluenesulphonic acid monohydrate, Sigma-Aldrich®
:
Merck
p-Toluenesulphonic acid monohydrate, Sigma-Aldrich®
:
Merck
:
6192-52-5
:
:
C₇H₈O₃S·1H₂O
:
MFCD00142137
:
190.22 g/mol
:
12,09671
:
102…104 °C
:
1.24 g/cm³ (20 °C)
:
180 °C
:
Ambient
:
203-180-0
:
2585
:
8,III
:
GHS07