Order Entry
Export
ContactUsLinkComponent
3,5-Dihydroxytoluene monohydrate
Irritant
3,5-Dihydroxytoluene monohydrate
Catalog # MOLE70074916-25G
Supplier:  Molekula
CAS Number:  
Irritant
3,5-Dihydroxytoluene monohydrate
Catalog # MOLE70074916-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :142.15 g/mol
Merck Index:12,06995
Boiling Pt:290 °C (1013 hPa)
Melting Pt:56…61 °C
Density:1.29 g/cm³ (4 °C)
Flash Pt:159 °C
Storage Temperature:Ambient
EINECS:207-984-2
 :GHS07
CAS Number:6153-39-5
Formula:C₇H₈O₂·1H₂O
MDL Number:MFCD00149092