
1-Octanesulphonic acid sodium salt monohydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207596-29-0
:
Formula:C₈H₁₇NaO₃S·1H₂O
MDL Number:MFCD00007544
:234.29 g/mol
Melting Pt:>300 °C
EINECS:226-195-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Loss on drying:7 % to 9 % (140°C)
- Titration after Ion exchange:≥97.5 %