

EDTA tetrasodium salt dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10378-23-1
:
Formula:C₁₀H₁₂N₂Na₄O₈·2H₂O
MDL Number:MFCD00150026
Merck Index:13,03544
Melting Pt:> 300 °C
:M-Clarity™ quality level = MQ200
Flash Pt:>100 °C
:416.2 g/mol
EINECS:200-573-9
:GHS05
:GHS07
Storage Temperature:Ambient