Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tri-Sodium citrate dihydrate
tri-Sodium citrate dihydrate, Sigma-Aldrich®
tri-Sodium citrate dihydrate, Sigma-Aldrich®
:
Merck
tri-Sodium citrate dihydrate, Sigma-Aldrich®
:
Merck
:
6132-04-3
:
:
Na₃C₆H₅O₇·2H₂O
:
MFCD00150031
:
294.1 g/mol
:
13,08675
:
150 °C
:
1.76 g/cm³ (20 °C)
:
Ambient
:
200-675-3