Trisodium citrate-5,5-hydrate, Sigma-Aldrich®
Catalog # SIAL25114-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:357.15 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-675-3
CAS Number:6858-44-2
Formula:C₆H₅Na₃O₇·5.5H₂O