
di-Ammonium oxalate monohydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:6009-70-7
:
Formula:(NH₄)₂C₂O₄·H₂O
MDL Number:MFCD00149694
:142.11 g/mol
Merck Index:13,00541
Boiling Pt:70 °C (1013 hPa)
Melting Pt:70 °C
Density:∼1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-202-3
UN:3288
ADR:6.1,III
:GHS07