di-Ammonium oxalate monohydrate
MOLE23230264-1KG
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
:
:142.11 g/mol
:13,00541
:70 °C (1013 hPa)
:70 °C
:∼1.5 g/cm³ (20 °C)
:Ambient
:214-202-3
:3288
:6.1,III
:GHS07
:6009-70-7
:(NH₄)₂C₂O₄·H₂O
:MFCD00149694