Manganese(II) acetate tetrahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:6156-78-1
:
Formula:(H₃CCOO)₂Mn·4H₂O
MDL Number:MFCD00062552
:245.09 g/mol
Merck Index:13,05746
Melting Pt:300 °C
Density:1.58 g/cm³ (20 °C)
EINECS:211-334-3