

Zinc acetate dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5970-45-6
:
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961
:219.51 g/mol
Merck Index:12,10256
Melting Pt:237 °C
Density:1.718 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-170-2
ADR:9,III
:M-Clarity™ quality level = MQ200
UN:3077
:GHS07
:GHS09