Lead(II) acetate trihydrate
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
- CAS Number:6080-56-4
- Formula:(H₃CCOO)₂Pb·3H₂O
- MDL Number:MFCD00150023
- Molecular Weight:379.34 g/mol
- Merck Index:13,05415
- Boiling Pt:280 °C (1013 hPa)
- Melting Pt:75 °C
- Density:2.55 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-104-4
- UN:1616
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
- CAS Number:6080-56-4
- Formula:(H₃CCOO)₂Pb·3H₂O
- MDL Number:MFCD00150023
- Molecular Weight:379.34 g/mol
- Merck Index:13,05415
- Boiling Pt:280 °C (1013 hPa)
- Melting Pt:75 °C
- Density:2.55 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-104-4
- UN:1616
- ADR:6.1,III



