Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium acetate trihydrate
Sodium acetate trihydrate, Sigma-Aldrich®
Sodium acetate trihydrate, Sigma-Aldrich®
:
Merck
Sodium acetate trihydrate, Sigma-Aldrich®
:
Merck
:
6131-90-4
:
:
H₃CCOONa·3H₂O
:
MFCD00071557
:
136.08 g/mol
:
13,08642
:
120…123 °C (1013 hPa)
:
58 °C
:
1.4 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ200
:
Ambient
:
204-823-8