Order Entry
Export
ContactUsLinkComponent
Silica gel drying agent, with moisture indicator cobalt (II) chloride (blue gel)
Health
Silica gel drying agent, with moisture indicator cobalt (II) chloride (blue gel)
Catalog # ROTH9351.1
Supplier:  Roth Carl
CAS Number:  
Health
Silica gel drying agent, with moisture indicator cobalt (II) chloride (blue gel)
Catalog # ROTH9351.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :60.08 g/mol
Melting Pt:1600 °C
 :GHS08
Storage Temperature:Ambient
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232