Order Entry
Export
ContactUsLinkComponent
N-(1-Naphthyl)ethylenediamine dihydrochloride
Irritant
N-(1-Naphthyl)ethylenediamine dihydrochloride
Catalog # ROTH4342.1
Supplier:  Roth Carl
CAS Number:  
undefined
Irritant
N-(1-Naphthyl)ethylenediamine dihydrochloride
Catalog # ROTH4342.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
 :259.18 g/mol
Merck Index:12,06496
Melting Pt:188…190 °C
EINECS:215-981-2
 :GHS07
Storage Temperature:Ambient
CAS Number:1465-25-4
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556