Hydroxylammonium chloride
Glass bottle for solids, 250 g
Catalog # ROTHP016.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Number:5470-11-1
- Formula:NH₂OH·HCl
- MDL Number:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:13,04853
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:151 °C
- Density:1.67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:226-798-2
- UN:2923
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



