Order Entry
Export
ContactUsLinkComponent
Guanidinium thiocyanate, Sigma-Aldrich®
Irritant
Guanidinium thiocyanate, Sigma-Aldrich®
Catalog # ROCH11685929001
Supplier:  ROCHE
CAS Number:  
Irritant
Guanidinium thiocyanate, Sigma-Aldrich®
Catalog # ROCH11685929001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :118.16 g/mol
Melting Pt:118 °C
Density:1.29 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:209-812-1
 :GHS07
Storage Temperature:Ambient
CAS Number:593-84-0
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027