Order Entry
Export
Orders LinkContactUsLinkComponent
Guanidine thiocyanate

Guanidine thiocyanate

Glass bottle for solids, 100 g
Catalog # MOLEM18449163
Irritant
Supplier:   Molekula
Guanidine thiocyanate
Catalog # MOLEM18449163
Supplier:  Molekula
CAS Number:  593-84-0
Guanidine thiocyanate

Glass bottle for solids, 100 g

CAS Number:  593-84-0
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Guanidinium sulphocyanide, GTC, Guanidium thiocyanate, Guanidinium rhodanide
  • CAS Number:593-84-0
  • Formula:CH₅N₃·CHNS
  • MDL Number:MFCD00013027
  • Molecular Weight:118.16 g/mol
  • Melting Pt:118 °C
  • Density:1.29 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-812-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications