ContactUsLinkComponent
Basic fuchsin
Basic fuchsin
  720-1070
 :  RAL
 :  
Basic fuchsin
  720-1070
 :  RAL
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
 :
 :337.85 g/mol
 :12,05686
 :235 °C
 :1.22 g/cm³ (20 °C)
 :211-189-6
 :GHS08
 :Ambient
 :1
 :632-99-5
 :58969-01-0
 :C₂₀H₂₀ClN₃
 :MFCD00012569