



Malachite green oxalate, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:2437-29-8
:18015-76-4
Formula:C₅₂H₅₄N₄O₁₂
MDL Number:MFCD00011766
:927.02 g/mol
Merck Index:13,05722
Melting Pt:∼159 °C
Storage Temperature:Room Temperature
EINECS:219-441-7
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS08
:GHS07
:GHS09
Specification Test Results
- Appearance (Colour):Metallic Green
Conforms - Appearance (Form):Powder or Solid or Crystals or Crystalline Chunk(s) or Chunk(s)
- Solubility (Colour)
1 mg/ml, H₂O:Dark Blue to Dark Green - Carbon:60.5 - 68.6%
- Nitrogen:5.3 - 6.6%
- UV-Vis Spectrum
in H₂O:616 - 620 nm