Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tetraethylammonium hydroxide
Tetraethylammonium hydroxide 25% in methanol, Sigma-Aldrich®
Tetraethylammonium hydroxide 25% in methanol, Sigma-Aldrich®
Tetraethylammonium hydroxide 25% in methanol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Tetraethylammonium hydroxide 25% in methanol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
77-98-5
:
Formula:
C₈H₂₁NO
MDL Number:
MFCD00009024
:
147.26 g/mol
EINECS:
201-073-3
UN:
2924
ADR:
3,II