Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrochloric acid 32%

Hydrochloric acid 32%

Corrosive
Irritant
Supplier:   Roth Carl
Hydrochloric acid 32%
Supplier:  Roth Carl
Certificates
Hydrochloric acid 32%

4 Options Available Below

Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:110 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:1.18 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 25,90 €
    25,90 €
    1 * 1 L
       25,90 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 23,80 €
    23,80 €
    1 * 1 L
       23,80 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 46,60 €
    46,60 €
    1 * 2,5 L
       46,60 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 44,60 €
    44,60 €
    1 * 2,5 L
       44,60 €