



Glutaraldehyde 25% (w/w) in water
:111-30-8
:
:C₅H₈O₂
:MFCD00007025
:100.12 g/mol
:101 °C (1013 hPa)
:–5 °C
:1.06 g/cm³
:≥ 71 °C
:203-856-5
:3265
:8,II
:GHS05
:GHS07
:GHS08
:GHS09
:Refrigerator
- Appearance:Clear solution
- Color scale:≤50 APHA
- Strength:24 to 26 %
- pH:3.2 to 4.2
- Methanol:≤0.3 % (GC)