(±)-trans-1,2-Cyclohexanediol 98%
:1460-57-7
:
:C₆H₁₂O₂
:MFCD00063611
:117 °C (17mbar)
:100…104 °C
:1,147 g/cm³ (24 °C)
:134 °C
:116.16 g/mol
:215-956-6
- Appearance:Off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:100 to 104 °C
- GC:≥97.5 %