Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-2-Methyl-2,4-pentanediol
(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®
(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®
:
Merck
(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®
:
Merck
:
107-41-5
:
:
C₆H₁₄O₂
:
MFCD00004547
:
118.18 g/mol
:
13,04730
:
198 °C (1013 hPa)
:
–40 °C
:
0.923 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ200
:
93 °C
:
203-489-0
:
GHS07
:
Ambient