(±)-3,7-Dimethyl-3-octanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-69-3
:
Formula:C₁₀H₂₂O
MDL Number:MFCD00004482
:158.28 g/mol
Boiling Pt:195.5…197 °C (1013 hPa)
Density:0.828 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:201-133-9