(±)-3,7-Dimethyl-3-octanol
Catalog # ROTH9446.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:158.28 g/mol
Boiling Pt:195.5…197 °C (1013 hPa)
Density:0.828 g/cm³ (20 °C)
EINECS:201-133-9
CAS Number:78-69-3
Formula:C₁₀H₂₂O
MDL Number:MFCD00004482