

3-Mercapto-1,2-propanediol ≥97%, BioXtra, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:96-27-5
:
Formula:C₃H₈O₂S
MDL Number:MFCD00004879
:108.16 g/mol
Merck Index:12,09471
Boiling Pt:118 °C/5 mmHg (lit.)
Density:1.25 g/mL (25 °C) (lit.)
:M-Clarity™ quality level = MQ200
EINECS:202-495-0
:GHS06
:GHS07
Storage Temperature:Refrigerator