
3-Mercapto-1,2-propanediol ≥98%
Catalog # ICNA0215572325
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Form:Clear Liquid
- Storage:Store at +4 °C, store under nitrogen
- Refractive Index:n20/D 1,527 (lit.)
Specification Test Results
About this item
:
:108.16 g/mol
Merck Index:12,09471
Boiling Pt:118 °C (1013 hPa)
Density:1.247 g/cm³ (20 °C)
EINECS:202-495-0
:GHS07
Storage Temperature:Ambient
CAS Number:96-27-5
Formula:C₃H₈O₂S
MDL Number:MFCD00004879