(±)-1,3-Butanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-88-0
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00004554
:90.12 g/mol
Merck Index:13,01566
Boiling Pt:206…208 °C (1013 hPa)
Melting Pt:–54 °C
Density:1.004…1.006 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:203-529-7