(±)-1,2-Pentanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5343-92-0
:
Formula:C₅H₁₂O₂
MDL Number:MFCD00010736
:104.15 g/mol
Boiling Pt:206 °C (1013 hPa)
Density:0.9802 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:226-285-3