
(±)-1,2-Hexanediol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6920-22-5
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00010737
:118.18 g/mol
Boiling Pt:223…224 °C
Melting Pt:45 °C
Density:0.951
Flash Pt:122 °C (251 °F)
EINECS:230-029-6
:GHS07
Storage Temperature:Ambient