Order Entry
Export
ContactUsLinkComponent
(±)-1,2-Decanediol, Sigma-Aldrich®
(±)-1,2-Decanediol, Sigma-Aldrich®
Catalog # SIAL260320-50G
Supplier:  Merck
CAS Number:  
(±)-1,2-Decanediol, Sigma-Aldrich®
Catalog # SIAL260320-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :174.28 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:214-288-2
CAS Number:1119-86-4
Formula:C₁₀H₂₂O₂
MDL Number:MFCD00010739