Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-1,2-Butanediol
(±)-1,2-Butanediol, Sigma-Aldrich®
(±)-1,2-Butanediol, Sigma-Aldrich®
:
Merck
(±)-1,2-Butanediol, Sigma-Aldrich®
:
Merck
:
584-03-2
:
26171-83-5
:
C₄H₁₀O₂
:
MFCD00004570
:
90.12 g/mol
:
193 °C (1013 hPa)
:
–50 °C
:
1.0189 g/cm³ (0 °C)
:
M-Clarity™ quality level = MQ200
:
209-527-2