Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Cyclohexadiene
1,4-Cyclohexadiene ≥97% stabilised
1,4-Cyclohexadiene ≥97% stabilised
:
Thermo Fisher Scientific
1,4-Cyclohexadiene ≥97% stabilised
:
Thermo Fisher Scientific
:
628-41-1
:
:
C₆H₈
:
MFCD00001535
:
88…90 °C
:
–49 °C
:
0.847
:
–6 °C (21 °F)
:
211-043-1
:
3295
:
3,II
:
Stabilised with hydroquinone (HQ) 0.10 %
:
80.13 g/mol
:
GHS02
:
GHS08
:
Refrigerator
msds_ALFAL07337
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years