1,4-Cyclohexadiene stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:628-41-1
:
Formula:C₆H₈
MDL Number:MFCD00001535
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–49.2 °C
Density:0.847 g/cm³ (20 °C)
Flash Pt:–10 °C
EINECS:211-043-1
ADR:3,II
:80.13 g/mol
UN:3295