Order Entry
Export
ContactUsLinkComponent
(±)-3-Amino-1,2-propanediol
(±)-3-Amino-1,2-propanediol
Catalog # MOLE47241522-25G
Supplier:  Molekula
CAS Number:  
(±)-3-Amino-1,2-propanediol
Catalog # MOLE47241522-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :91.11 g/mol
Boiling Pt:264 °C (1013 hPa)
Density:1.1752 g/cm³ (20 °C)
EINECS:210-475-8
CAS Number:616-30-8
Formula:C₃H₉NO₂
MDL Number:MFCD00008140