(±)-3-Amino-1,2-propanediol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:616-30-8
:
Formula:C₃H₉NO₂
MDL Number:MFCD00008140
:91.11 g/mol
Boiling Pt:264…265 °C (739 mmHg)
Density:1.17 g/cm³
Flash Pt:155 °C
Storage Temperature:Ambient
EINECS:210-475-8
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4900 to 1.4940 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- Titration with HClO4:≥97.5 %