3-Phenoxypropionic acid
Glass bottle for solids, 5 g
Catalog # MOLEM29353362
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3-Penoxypropionic acid, 3-Phenoxypropanoic acid
- CAS Number:7170-38-9
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00002769
- Molecular Weight:166.18 g/mol
- Boiling Pt:235 °C (1013 hPa)
- Melting Pt:92 °C
- EINECS:230-518-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



