Order Entry
Export
Orders LinkContactUsLinkComponent
3-Phenoxypropionic acid ≥98%

3-Phenoxypropionic acid ≥98%

3-Phenoxypropionic acid ≥98%
Certificates
3-Phenoxypropionic acid ≥98%
3-Phenoxypropionic acid ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 7170-38-9 Molecular Formula: C6H5O(CH2)2CO2H Formula Weight: 166.17 Melting Point: 96-99° Boiling Point: 243-245° MDL No.: MFCD00002769

Synonyms:  3-Penoxypropionic acid, 3-Phenoxypropanoic acid
  • CAS Number:7170-38-9
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002769
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:243…245 °C
  • Melting Pt:96…99 °C
  • Storage Temperature:Ambient
  • EINECS:230-518-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,90 €
    21,90 €
    1 * 1 g
       21,90 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 57,10 €
    57,10 €
    1 * 5 g
       57,10 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 171,00 €
    171,00 €
    1 * 25 g
       171,00 €