Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Diiodopentane
1,5-Diiodopentane ≥97% stabilised
1,5-Diiodopentane ≥97% stabilised
:
Thermo Fisher Scientific
1,5-Diiodopentane ≥97% stabilised
:
Thermo Fisher Scientific
:
628-77-3
:
:
C₅H₁₀I₂
:
MFCD00001101
:
323.94 g/mol
:
128…130 °C (10 mmHg)
:
9 °C
:
2.176
:
>110 °C (230 °F)
:
Stabilised with copper
:
211-054-1
:
GHS07
:
Ambient
msds_ALFAA17298
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years